N-苯基乙二胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
N-(2-氨基乙基)苯胺
P1101-10G
P1101-25G
TCIAP1101-10GEA
0
CNY
TCIAP1101-10G
TCIAP1101-25G
N-苯基乙二胺 ≥98.0% (by GC, titration analysis)
N-苯基乙二胺
分子式:
C₆H₅NHCH₂CH₂NH₂ 分子量: 136.2 g/mol 沸点: 263°C 闪点: 113 |
MDL编号:
MFCD00008162 CAS号: 1664-40-0 EINECS: 216-773-4 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific gravity (20/20) | 1.0460 to 1.0480 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...