三(4-甲氧基苯基)膦 ≥97.0% (通过滴定分析)
供应商: TCI
|
Warning
|
T0861-25G
T0861-5G
TCIAT0861-25GEA
0
CNY
TCIAT0861-25G
TCIAT0861-5G
三(4-甲氧基苯基)膦 ≥97.0% (通过滴定分析)
三(4-甲氧基苯基)膦
分子式:
C₂₁H₂₁O₃P 沸点: 446 °C (1013 hPa) 溶点: 135 °C |
MDL编号:
MFCD00014896 CAS号: 855-38-9 EINECS: 212-723-0 |
|
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Melting point | 130.0 to 135.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...