反式-2-己烯酸 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
trans-Hex-2-enoic acid
H0383-25G
H0383-250G
TCIAH0383-25GEA
0
CNY
TCIAH0383-25G
TCIAH0383-250G
反式-2-己烯酸 ≥98.0% (by GC)
反式-2-己烯酸
分子式:
C₆H₁₀O₂ 分子量: 114.14 g/mol 沸点: 118°C 溶点: 32 密度: 0.946 |
MDL编号:
MFCD00002705 CAS号: 13419-69-7 EINECS: 236-528-5 UN号: 3261 ADR: 8 III |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...