四苯基环戊二烯酮 ≥98.0% (by HPLC)
供应商: TCI
Synonyms:
2,3,4,5-Tetraphenylcyclopenta-2,4-dienone
T1062-25G
T1062-5G
TCIAT1062-25GEA
0
CNY
TCIAT1062-25G
TCIAT1062-5G
四苯基环戊二烯酮 ≥98.0% (by HPLC)
四苯基环戊二烯酮
分子式:
C₂₉H₂₀O 溶点: 219…222 °C 密度: 1,233 g/cm³ (20 °C) |
MDL编号:
MFCD00001407 CAS号: 479-33-4 EINECS: 207-530-3 |
|
产品技术规格
Appearance | Black powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...