甲酸戊酯 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Formic acid amyl ester, Formic acid pentyl ester, Amyl formate
F0049-25ML
F0049-500ML
TCIAF0049-25MLEA
0
CNY
TCIAF0049-25ML
TCIAF0049-500ML
甲酸戊酯 ≥95.0% (by GC)
甲酸戊酯
分子式:
C₆H₁₂O₂ 沸点: 130°C 溶点: –73 °C 密度: 0,89 g/cm³ (20 °C) 闪点: 32°C |
MDL编号:
MFCD00040440 CAS号: 638-49-3 EINECS: 211-340-6 UN号: 1109 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...