甲酸己酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Formic acid hexyl ester, n-Hexyl formate
H1271-25ML
H1271-500ML
TCIAH1271-25MLEA
0
CNY
TCIAH1271-25ML
TCIAH1271-500ML
甲酸己酯 ≥98.0% (by GC)
甲酸己酯
分子式:
C₇H₁₄O₂ 沸点: 156°C 溶点: –63°C 密度: 0,875 g/cm³ (20 °C) 闪点: 48°C |
MDL编号:
MFCD00046146 CAS号: 629-33-4 EINECS: 211-087-1 UN号: 3272 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...