甲酸苄酯 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: Formic acid benzyl ester

F0050-25G F0050-500G
TCIAF0050-25GEA 0 CNY
TCIAF0050-25G TCIAF0050-500G
甲酸苄酯 ≥95.0% (by GC)
甲酸苄酯
分子式: C₈H₈O₂
分子量: 136.15 g/mol
沸点: 201°C
闪点: 94°C
MDL编号: MFCD00014128
CAS号: 104-57-4
EINECS: 203-214-4
Merck Index: 14,01134

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR