氰乙酸叔丁酯 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
tert-Butyl 2-cyanoacetate
C1441-25ML
C1441-250ML
TCIAC1441-25MLEA
0
CNY
TCIAC1441-25ML
TCIAC1441-250ML
氰乙酸叔丁酯 ≥97.0% (by GC)
氰乙酸叔丁酯
分子式:
C₇H₁₁NO₂ 分子量: 141.17 g/mol 沸点: 108°C 闪点: 91°C 存储条件: Refrigerator |
MDL编号:
MFCD00001938 CAS号: 1116-98-9 EINECS: 214-243-7 |
|
产品技术规格
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...