油醇 >60.0% (通过GC)
供应商: TCI
|
Warning
|
O0058-25ML
O0058-500ML
TCIAO0058-25MLEA
0
CNY
TCIAO0058-25ML
TCIAO0058-500ML
油醇 >60.0% (通过GC)
油醇
分子式:
C₁₈H₃₆O 沸点: 207°C 溶点: 13…19 °C 密度: 0,85 g/cm³ (20 °C) 闪点: 189°C |
MDL编号:
MFCD00002993 CAS号: 143-28-2 EINECS: 205-597-3 Merck Index: 14,06831 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 60.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...