N-乙基马来酰亚胺 ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
|
Danger
|
Synonyms:
NEM
E0136-1G
E0136-25G
E0136-5G
TCIAE0136-1GEA
0
CNY
TCIAE0136-1G
TCIAE0136-25G
TCIAE0136-5G
N-乙基马来酰亚胺 ≥98.0% (by HPLC, total nitrogen)
N-乙基马来酰亚胺
分子式:
C₆H₇NO₂ 分子量: 125.13 g/mol 沸点: 210°C 溶点: 46 存储条件: Refrigerator |
MDL编号:
MFCD00005509 CAS号: 128-53-0 EINECS: 204-892-4 UN号: 2928 ADR: 6.1 II Merck Index: 14,03822 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 44.0 to 47.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...