6-氯-1-己炔 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 6-Chlorohex-1-yne

C1493-5ML C1493-25ML
TCIAC1493-5MLEA 0 CNY
TCIAC1493-5ML TCIAC1493-25ML
6-氯-1-己炔 ≥97.0% (by GC)
6-氯-1-己炔
分子式: C₆H₉Cl
分子量: 116.59 g/mol
沸点: 144°C
闪点: 34°C
MDL编号: MFCD00013697
CAS号: 10297-06-0
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4ba5d952a73d026ef0308a9917979ac93432e33&vid=e4ba5d952a73d026ef0308a9917979ac93432e33&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR