5-甲基噻唑 ≥98.0% (通过GC)

供应商: TCI
Warning

M2234-25G M2234-5G
TCIAM2234-25GEA 0 CNY
TCIAM2234-25G TCIAM2234-5G
5-甲基噻唑 ≥98.0% (通过GC)
5-甲基噻唑
分子式: C₄H₅NS
沸点: 142°C
密度: 1,127 g/cm³ (20 °C)
闪点: 42°C
MDL编号: MFCD00005335
CAS号: 3581-89-3
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67c1f2a8bfaa79744fb7913a3f80b55dd86136cd&vid=67c1f2a8bfaa79744fb7913a3f80b55dd86136cd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR