2-碘芴 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-碘-9H-芴

I0793-5G I0793-25G
TCIAI0793-5GEA 0 CNY
TCIAI0793-5G TCIAI0793-25G
2-碘芴 ≥95.0% (by GC)
2-碘芴
分子式: C₁₃H₉I
MDL编号: MFCD00019049
CAS号: 2523-42-4
UN号: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6778080ad256bfabc19d125940fb833da7871452&vid=6778080ad256bfabc19d125940fb833da7871452&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR