乙酰丙酸甲酯 ≥99.0% (by GC)

供应商: TCI

Synonyms: 4-Ketovaleric acid methyl ester, 4-Oxopentanoic acid methyl ester, Methyl 4-oxopentanoate, 4-Oxovaleric acid methyl ester, Methyl 4-oxovalerate, Levulinic acid methyl ester

L0044-25G L0044-500G
TCIAL0044-25GEA 0 CNY
TCIAL0044-25G TCIAL0044-500G
乙酰丙酸甲酯 ≥99.0% (by GC)
乙酰丙酸甲酯
分子式: C₆H₁₀O₃
分子量: 130.14 g/mol
沸点: 196°C
溶点: -24
闪点: 72°C
MDL编号: MFCD00017499
CAS号: 624-45-3
EINECS: 210-846-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4767e67abcd22e45b6e2a3c2db040d5f97065b7&vid=b4767e67abcd22e45b6e2a3c2db040d5f97065b7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR