4-甲基噻唑 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
M0894-25G
M0894-5G
TCIAM0894-25GEA
0
CNY
TCIAM0894-25G
TCIAM0894-5G
4-甲基噻唑 ≥98.0% (通过GC)
4-甲基噻唑
分子式:
C₄H₅NS 沸点: 134°C 密度: 1,1187 g/cm³ (20 °C) 闪点: 32°C |
MDL编号:
MFCD00005340 CAS号: 693-95-8 EINECS: 211-764-1 UN号: 1993 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...