对甲基苯甲酸甲酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Methylbenzoic acid methyl ester, Methyl-4-methylbenzoate, p-Toluic acid methyl ester, 4-Toluic acid methyl ester, Methyl 4-methylbenzoate
T0297-25G
T0297-500G
TCIAT0297-25GEA
0
CNY
TCIAT0297-25G
TCIAT0297-500G
对甲基苯甲酸甲酯 ≥98.0% (by GC)
对甲基苯甲酸甲酯
分子式:
C₉H₁₀O₂ 分子量: 150.18 沸点: 217 密度: 1.06 闪点: 89 |
MDL编号:
MFCD00008441 CAS号: 99-75-2 EINECS: 202-784-1 |
|
产品技术规格
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...