Heptyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: Heptylparaben, n-Heptyl 4-hydroxybenzoate, Heptyl parahydroxybenzoate, 4-Hydroxybenzoic acid heptyl ester

H0212-25G H0212-500G
TCIAH0212-25GEA 0 CNY
TCIAH0212-25G TCIAH0212-500G
Heptyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
Heptyl-4-hydroxybenzoate
分子式: C₁₄H₂₀O₃
分子量: 236.31 g/mol
存储条件: Ambient
MDL编号: MFCD00016481
CAS号: 1085-12-7
EINECS: 214-115-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f98e2993ed292483ae5ff759f4627ad1fb165d4f&vid=f98e2993ed292483ae5ff759f4627ad1fb165d4f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 45.0 to 50.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR