Dodecyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
Dodecylparaben, Dodecyl parahydroxybenzoate, 4-Hydroxybenzoic acid dodecyl ester
H0352-25G
H0352-500G
TCIAH0352-25GEA
0
CNY
TCIAH0352-25G
TCIAH0352-500G
Dodecyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
Dodecyl-4-hydroxybenzoate
分子式:
C₁₉H₃₀O₃ 分子量: 306.45 g/mol 存储条件: Ambient |
MDL编号:
MFCD00016473 CAS号: 2664-60-0 EINECS: 220-195-8 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 52.0 to 58.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...