4-Aminophthalimide ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
5-Aminoisoindoline-1,3-dione, 5-Amino-isoindole-1,3-dione
A0966-25G
A0966-5G
TCIAA0966-25GEA
0
CNY
TCIAA0966-25G
TCIAA0966-5G
4-Aminophthalimide ≥98.0% (by GC)
4-Aminophthalimide
分子式:
C₈H₆N₂O₂ 沸点: 447 °C (1013 hPa) 溶点: 294 °C |
MDL编号:
MFCD00041854 CAS号: 3676-85-5 EINECS: 222-948-6 |
|
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in N,N-DMF | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...