3-甲基苯并呋喃 ≥98.0% (by GC)

供应商: TCI

M2112-1G M2112-5G
TCIAM2112-1GEA 0 CNY
TCIAM2112-1G TCIAM2112-5G
3-甲基苯并呋喃 ≥98.0% (by GC)
3-甲基苯并呋喃
分子式: C₉H₈O
分子量: 132.16 g/mol
沸点: 197°C
溶点: 19
密度: 1.0568
存储条件: Refrigerator
MDL编号: MFCD00205641
CAS号: 21535-97-7
EINECS: 244-427-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fb330d151a8e04bc9d788fdc5c5dd9ae3356586&vid=8fb330d151a8e04bc9d788fdc5c5dd9ae3356586&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR