Ursodeoxycholic acid ≥98.0% (通过滴定分析)

供应商: TCI

U0030-25G U0030-5G
TCIAU0030-25GEA 0 CNY
TCIAU0030-25G TCIAU0030-5G
Ursodeoxycholic acid ≥98.0% (通过滴定分析)
Ursodeoxycholic acid
分子式: C₂₄H₄₀O₄
沸点: 547 °C (1013 hPa)
溶点: 205 °C
MDL编号: MFCD00003680
CAS号: 128-13-2
EINECS: 204-879-3
Merck Index: 14,09889
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1571318212d73f19547c9fd788b5df8ed0807c7e&vid=1571318212d73f19547c9fd788b5df8ed0807c7e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 98.0 % (after drying)
Melting point 200.0 to 205.0 deg-C
Specific rotation [a]20/D + 59.0 to + 62.0 deg (C=4, EtOH)
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR