3,5-二甲基异噁唑 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
D1767-25ML
D1767-250ML
TCIAD1767-25MLEA
0
CNY
TCIAD1767-25ML
TCIAD1767-250ML
3,5-二甲基异噁唑 ≥99.0% (by GC)
3,5-二甲基异噁唑
分子式:
C₅H₇NO 沸点: 141°C 密度: 0,98 g/cm³ (20 °C) 闪点: 31°C |
MDL编号:
MFCD00003156 CAS号: 300-87-8 EINECS: 206-100-2 UN号: 1993 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...