3,5-叔丁基苯甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
D2663-1G
D2663-25G
D2663-5G
TCIAD2663-1GEA
0
CNY
TCIAD2663-1G
TCIAD2663-25G
TCIAD2663-5G
3,5-叔丁基苯甲酸 ≥98.0% (by GC, titration analysis)
3,5-叔丁基苯甲酸
分子式:
C₁₅H₂₂O₂ |
MDL编号:
MFCD00082727 CAS号: 16225-26-6 EINECS: 240-350-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...