3-吡啶甲醇 ≥98.0% (通过GC)

供应商: TCI
Warning

P0555-25ML P0555-500ML
TCIAP0555-25MLEA 0 CNY
TCIAP0555-25ML TCIAP0555-500ML
3-吡啶甲醇 ≥98.0% (通过GC)
3-吡啶甲醇
分子式: C₆H₇NO
沸点: 145°C
溶点: –7 °C
密度: 1,1357 g/cm³ (20 °C)
闪点: 157°C
MDL编号: MFCD00006407
CAS号: 100-55-0
EINECS: 202-864-6
Merck Index: 14,06527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac4814d0293bc015ae762accca3949c5057e6f5a&vid=ac4814d0293bc015ae762accca3949c5057e6f5a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR