3-吡啶甲基胺 ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: 3-氨甲基吡啶

P0762-25ML P0762-500ML
TCIAP0762-25MLEA 0 CNY
TCIAP0762-25ML TCIAP0762-500ML
3-吡啶甲基胺 ≥99.0% (by GC)
3-吡啶甲基胺
分子式: C₆H₈N₂
分子量: 108.14 g/mol
沸点: 74°C
溶点: -21
闪点: 100°C
MDL编号: MFCD00006412
CAS号: 3731-52-0
EINECS: 223-091-0
UN号: 2735
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebacb546cd22dad73fc4a1e4f7f84a13bee8e319&vid=ebacb546cd22dad73fc4a1e4f7f84a13bee8e319&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR