3-(2-Aminoethyl)pyridine ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2-(Pyridin-3-yl)ethanamine, 2-(3-Pyridyl)ethylamine

A1336-25G A1336-5G
TCIAA1336-25GEA 0 CNY
TCIAA1336-25G TCIAA1336-5G
3-(2-Aminoethyl)pyridine ≥98.0% (by GC, titration analysis)
3-(2-Aminoethyl)pyridine
分子式: C₇H₁₀N₂
分子量: 122.17 g/mol
沸点: 128°C
MDL编号: MFCD00191604
CAS号: 20173-24-4
UN号: 2735
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=385806887e683cb14acbd76697c92dda58d57473&vid=385806887e683cb14acbd76697c92dda58d57473&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR