邻甲基苯甲酸甲酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 邻甲苯甲酸甲酯

T0296-25ML T0296-500ML
TCIAT0296-25MLEA 0 CNY
TCIAT0296-25ML TCIAT0296-500ML
邻甲基苯甲酸甲酯 ≥98.0% (by GC)
邻甲基苯甲酸甲酯
分子式: C₉H₁₀O₂
分子量: 150.18
沸点: 213°C
溶点: -50
密度: 1.079
闪点: 82°C
MDL编号: MFCD00008428
CAS号: 89-71-4
EINECS: 201-932-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR