2-甲氧基噻唑 ≥98.0% (通过GC)

供应商: TCI
Warning

M1753-25G M1753-5G
TCIAM1753-25GEA 0 CNY
TCIAM1753-25G TCIAM1753-5G
2-甲氧基噻唑 ≥98.0% (通过GC)
2-甲氧基噻唑
分子式: C₄H₅NOS
沸点: 150°C
MDL编号: MFCD01631143
CAS号: 14542-13-3
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc7f9a1867ec9c0b4fc8cb8691b3c67e6c4998e5&vid=bc7f9a1867ec9c0b4fc8cb8691b3c67e6c4998e5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR