2-乙基-4-甲基咪唑 ≥95.0% (by GC)

供应商: TCI
Danger

Synonyms: 2-Ethyl-4-methyl-1H-imidazole

E0232-25G E0232-500G
TCIAE0232-25GEA 0 CNY
TCIAE0232-25G TCIAE0232-500G
2-乙基-4-甲基咪唑 ≥95.0% (by GC)
2-乙基-4-甲基咪唑
分子式: C₆H₁₀N₂
分子量: 110.16 g/mol
沸点: 292
溶点: 45
MDL编号: MFCD00005193
CAS号: 931-36-2
EINECS: 213-234-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51200c50ceba030f315e624ea1210e180646597c&vid=f3c95c6a1178bf7c7fc9654779608611add5dfdc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR