2-乙酰基-5-氯噻酚 ≥99.0% (by GC)
供应商: TCI
Synonyms: 5-氯-2-噻吩基甲基酮
A1471-25G A1471-5G
TCIAA1471-25GEA 0 CNY
TCIAA1471-25G TCIAA1471-5G
2-乙酰基-5-氯噻酚 ≥99.0% (by GC)
2-乙酰基-5-氯噻酚
分子式: C₆H₅ClOS
沸点: 118°C
溶点: 52 °C
MDL编号: MFCD00005444
CAS号: 6310-09-4
EINECS: 228-630-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=480be349890f1b331ca30e5962308f7af040ee45&vid=480be349890f1b331ca30e5962308f7af040ee45&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 45.0 to 48.0 deg-C