2-溴-3-(三氟甲基)吡啶 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
B3179-1G
B3179-5G
TCIAB3179-1GEA
0
CNY
TCIAB3179-1G
TCIAB3179-5G
2-溴-3-(三氟甲基)吡啶 ≥98.0% (by GC)
2-溴-3-(三氟甲基)吡啶
分子式:
C₆H₃BrF₃N 分子量: 226 g/mol 沸点: 69°C |
MDL编号:
MFCD00153084 CAS号: 175205-82-0 UN号: 2811 ADR: 6.1 II |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...