2-Amino-5-phenylpyridine ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-Phenylpyridin-2-amine, 5-Phenyl-2-pyridylamine
A1667-1G
A1667-100MG
TCIAA1667-1GEA
0
CNY
TCIAA1667-1G
TCIAA1667-100MG
2-Amino-5-phenylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-phenylpyridine
分子式:
C₁₁H₁₀N₂ 溶点: 135°C |
MDL编号:
MFCD01692452 CAS号: 33421-40-8 |
|
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 134.0 to 138.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...