2-氨基-4-(三氟甲基)吡啶 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-(Trifluoromethyl)pyridin-2-amine, 4-(Trifluoromethyl)-2-pyridylamine

A2168-1G A2168-200MG A2168-5G
TCIAA2168-1GEA 0 CNY
TCIAA2168-1G TCIAA2168-200MG TCIAA2168-5G
2-氨基-4-(三氟甲基)吡啶 ≥98.0% (by GC, titration analysis)
2-氨基-4-(三氟甲基)吡啶
分子式: C₆H₅F₃N₂
分子量: 162.11 g/mol
MDL编号: MFCD00077455
CAS号: 106447-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f1bc11b554d1b99a39413cb88b1637e32067ba0&vid=0f1bc11b554d1b99a39413cb88b1637e32067ba0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR