2-氨基-2-甲基-1,3-丙二醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
AMPD, 氨甲基丙二醇
A0332-25G
A0332-500G
TCIAA0332-25GEA
0
CNY
TCIAA0332-25G
TCIAA0332-500G
2-氨基-2-甲基-1,3-丙二醇 ≥98.0% (by GC, titration analysis)
2-氨基-2-甲基-1,3-丙二醇
分子式:
C₄H₁₁NO₂ 分子量: 105.14 g/mol 沸点: 272 |
MDL编号:
MFCD00004678 CAS号: 115-69-5 EINECS: 204-100-7 Merck Index: 13,00447 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 106.0 to 111.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...