2,5-二氯-3-乙酰基噻吩 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,5-二氯-3-噻吩基甲基酮

A1182-25G A1182-5G
TCIAA1182-25GEA 0 CNY
TCIAA1182-25G TCIAA1182-5G
2,5-二氯-3-乙酰基噻吩 ≥98.0% (by GC)
2,5-二氯-3-乙酰基噻吩
分子式: C₆H₄Cl₂OS
分子量: 195.07 g/mol
沸点: 122°C
MDL编号: MFCD00014522
CAS号: 36157-40-1
EINECS: 252-893-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7e15eef51142d38114688d0bfd928e0ab5c6d4f&vid=f7e15eef51142d38114688d0bfd928e0ab5c6d4f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystalline
Purity (GC) min. 98.0 %
Melting point 38.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR