2,4-二甲基噻唑 ≥99.0% (by GC)

供应商: TCI
Warning

D1220-25ML D1220-5ML
TCIAD1220-25MLEA 0 CNY
TCIAD1220-25ML TCIAD1220-5ML
2,4-二甲基噻唑 ≥99.0% (by GC)
2,4-二甲基噻唑
分子式: C₅H₇NS
沸点: 144 °C (1013 hPa)
密度: 1,0601 g/cm³ (15 °C)
闪点: 43°C
MDL编号: MFCD00014509
CAS号: 541-58-2
EINECS: 208-786-9
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=942538c4618415ca00364078e36134ae24493e54&vid=942538c4618415ca00364078e36134ae24493e54&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR