5-乙基脲 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
2,4-二羟基-5-乙基嘧啶, 5-乙基尿嘧啶
E0807-1G
E0807-5G
TCIAE0807-1GEA
0
CNY
TCIAE0807-1G
TCIAE0807-5G
5-乙基脲 ≥98.0% (by HPLC, titration analysis)
5-乙基脲
分子式:
C₆H₈N₂O₂ 溶点: 303°C |
MDL编号:
MFCD00079187 CAS号: 4212-49-1 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...