2,4-二溴噻唑 ≥98.0% (通过GC)

供应商: TCI
Warning

D3667-1G D3667-25G D3667-5G
TCIAD3667-1GEA 0 CNY
TCIAD3667-1G TCIAD3667-25G TCIAD3667-5G
2,4-二溴噻唑 ≥98.0% (通过GC)
2,4-二溴噻唑
分子式: C₃HBr₂NS
MDL编号: MFCD01318994
CAS号: 4175-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481765d28b80430d26fc534212e7dd91ea02588c&vid=481765d28b80430d26fc534212e7dd91ea02588c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR