2,4,5-三苯基咪唑 ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 咯吩

T0999-25G T0999-5G
TCIAT0999-25GEA 0 CNY
TCIAT0999-25G TCIAT0999-5G
2,4,5-三苯基咪唑 ≥98.0% (by HPLC, titration analysis)
2,4,5-三苯基咪唑
分子式: C₂₁H₁₆N₂
分子量: 296.37 g/mol
沸点: 509
溶点: 270
MDL编号: MFCD00005187
CAS号: 484-47-9
EINECS: 207-606-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa1caa998c1c82597a66a909b90ca77dba7f9bc2&vid=fa1caa998c1c82597a66a909b90ca77dba7f9bc2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR