2,3-二甲基呋喃 ≥98.0% (by GC)

供应商: TCI
Danger

D2865-1G D2865-5G
TCIAD2865-1GEA 0 CNY
TCIAD2865-1G TCIAD2865-5G
2,3-二甲基呋喃 ≥98.0% (by GC)
2,3-二甲基呋喃
分子式: C₆H₈O
沸点: 92 - 94 °C (1013 hPa)
密度: 0,9285 g/cm³ (20 °C)
闪点: –1°C
存储条件: Refrigerator
MDL编号: MFCD00153893
CAS号: 14920-89-9
UN号: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=331965e309c1bd7e05e25087512d0395fd4b79ea&vid=331965e309c1bd7e05e25087512d0395fd4b79ea&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR