1-(2-吡啶基)哌嗪 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-哌嗪吡啶
P1033-25G
P1033-5G
TCIAP1033-25GEA
0
CNY
TCIAP1033-25G
TCIAP1033-5G
1-(2-吡啶基)哌嗪 ≥98.0% (by GC, titration analysis)
1-(2-吡啶基)哌嗪
分子式:
C₉H₁₃N₃ 沸点: 122°C 溶点: 85…87 °C 密度: 1,07 g/cm³ (20 °C) |
MDL编号:
MFCD00006216 CAS号: 34803-66-2 EINECS: 252-220-3 |
|
产品技术规格
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...