N-Phenylsuccinimide ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
N-PhenylsucciniMide
P1359-25G
P1359-5G
TCIAP1359-25GEA
0
CNY
TCIAP1359-25G
TCIAP1359-5G
N-Phenylsuccinimide ≥98.0% (by GC)
N-Phenylsuccinimide
分子式:
C₁₀H₉NO₂ 存储条件: Refrigerator |
MDL编号:
MFCD00022587 CAS号: 83-25-0 Merck Index: 14,08867 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...