N-甲基琥珀酰亚胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Methylpyrrolidine-2,5-dione
M2207-5G
M2207-25G
TCIAM2207-5GEA
0
CNY
TCIAM2207-5G
TCIAM2207-25G
N-甲基琥珀酰亚胺 ≥98.0% (by GC)
N-甲基琥珀酰亚胺
分子式:
C₅H₇NO₂ 沸点: 235°C 溶点: 66…69 °C 密度: 1,37 g/cm³ (20.85 °C) |
MDL编号:
MFCD00005517 CAS号: 1121-07-9 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...