4,4'-二甲氧基二苯甲醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
双(4-甲氧苯基)甲醇
D2141-25G
D2141-5G
TCIAD2141-25GEA
0
CNY
TCIAD2141-25G
TCIAD2141-5G
4,4'-二甲氧基二苯甲醇 ≥98.0% (by GC)
4,4'-二甲氧基二苯甲醇
分子式:
C₁₅H₁₆O₃ 分子量: 244.29 g/mol 沸点: 362 |
MDL编号:
MFCD00008410 CAS号: 728-87-0 EINECS: 211-975-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...