2-氟苯乙醇 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Fluorophenethyl alcohol
F0778-25G
F0778-5G
TCIAF0778-25GEA
0
CNY
TCIAF0778-25G
TCIAF0778-5G
2-氟苯乙醇 ≥95.0% (by GC)
2-氟苯乙醇
分子式:
C₈H₉FO 分子量: 140.16 g/mol 沸点: 137°C 闪点: 98°C |
MDL编号:
MFCD00002887 CAS号: 50919-06-7 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...