1-Hexyl-4-ethynylbenzene ≥95.0% (by GC)
供应商: TCI
Synonyms:
4-Hexylphenylacetylene, 1-Ethynyl-4-hexylbenzene, (4-Hexylphenyl)acetylene, 4-Hexyl-1-ethynylbenzene, (4-Hexylphenyl)ethyne
E0564-25G
E0564-5G
TCIAE0564-25GEA
0
CNY
TCIAE0564-25G
TCIAE0564-5G
1-Hexyl-4-ethynylbenzene ≥95.0% (by GC)
1-Hexyl-4-ethynylbenzene
分子式:
C₁₄H₁₈ |
MDL编号:
MFCD00173883 CAS号: 79887-11-9 |
|
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...