3,5-Bis(trifluoromethyl)benzonitrile ≥98.0% (通过GC)

供应商: TCI

B1896-25G B1896-5G
TCIAB1896-25GEA 0 CNY
TCIAB1896-25G TCIAB1896-5G
3,5-Bis(trifluoromethyl)benzonitrile ≥98.0% (通过GC)
3,5-Bis(trifluoromethyl)benzonitrile

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 19.0 to 23.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR