2-(三氟甲基)苯甲腈 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
α,α,α-三氟邻甲基苯甲腈
C1004-25G
C1004-5G
TCIAC1004-25GEA
0
CNY
TCIAC1004-25G
TCIAC1004-5G
2-(三氟甲基)苯甲腈 ≥98.0% (by GC)
2-(三氟甲基)苯甲腈
分子式:
C₈H₄F₃N 分子量: 171.12 g/mol 沸点: 206°C 溶点: 18 闪点: 90°C |
MDL编号:
MFCD00001791 CAS号: 447-60-9 EINECS: 207-184-3 UN号: 3276 ADR: 6.1 III |
|
产品技术规格
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...