4-氯苄硫醇 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 4-氯苯甲基硫醇

C0658-25G C0658-500G
TCIAC0658-25GEA 0 CNY
TCIAC0658-25G TCIAC0658-500G
4-氯苄硫醇 ≥98.0% (by GC)
4-氯苄硫醇
分子式: C₇H₇ClS
分子量: 158.65
沸点: 103°C
密度: 1.2
闪点: 76°C
MDL编号: MFCD00004870
CAS号: 6258-66-8
EINECS: 228-395-7
UN号: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e76176cc02bffc4b4ff1852e409d1c61abd7fb1&vid=2e76176cc02bffc4b4ff1852e409d1c61abd7fb1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR