4-氯-2-氟碘苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
5-Chloro-1-fluoro-2-iodobenzene, 4-Chloro-2-fluoro-1-iodobenzene, 4-Chloro-2-fluoroiodobenzene
C1499-25G
C1499-5G
TCIAC1499-25GEA
0
CNY
TCIAC1499-25G
TCIAC1499-5G
4-氯-2-氟碘苯 ≥98.0% (by GC)
4-氯-2-氟碘苯
分子式:
C₆H₃ClFI 分子量: 256.45 g/mol |
MDL编号:
MFCD00079729 CAS号: 6797-79-1 |
|
产品技术规格
Appearance | White or Colorles to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...